Learn More
SERVALYT™ 9-11 Carrier Ampholytes, Serva Electrophoresis™ Product Code.: 11777998
Estimated Shipment: 27-05-2025
Add to basket
SERVALYT™ 9-11 Carrier Ampholytes, Serva Electrophoresis™
Product Code. 11777998
10 mL
Encompass_Preferred
Click to view available options
Quantity:
10 mL
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11777998
Brand: Serva Electrophoresis 4290901
Estimated Shipment: 27-05-2025
Add to basket
Add to basket
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Specifications
Physical Form Liquid
Composition SERVALYT 9-11
Color Brown
Quantity 10 mL
Odor Uncharacteristic
Solubility Information Fully miscible
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
SERVALYT™ 9-11 Carrier Ampholytes, Serva Electrophoresis™