Learn More
Interscience™ Set of Tygon™ Tubings for Pump Head
20 cm long Tygon™ tubings for use with DiluFlow and FlexiPump pump heads.
63.30€
Specifications
Product Type | Pump Tubing |
---|---|
Autoclavable | Autoclavable |
For Use With (Application) | Adding Tubing to Dilutor Pumps |
Quantity | 6 Tubings/Set |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
12818291
|
Interscience™
513011 |
DiluFlow, DiluFlow Pro, DiluFlow Elite (5 kg), FlexiPump, FlexiPump Pro |
63.30€
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
17860087
|
Interscience™
561304 |
DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro |
63.30€
Pack of 6 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
Description
- Transparent
- Flexible Tygon™ tubing
Specifications
Pump Tubing | |
Autoclavable | |
Adding Tubing to Dilutor Pumps | |
6 Tubings/Set |
Product Title
Interscience™ Set of Tygon™ Tubings for Pump Head