missing translation for 'onlineSavingsMsg'
Learn More

SF3A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

459.00€

Specifications

Antigen SF3A2
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18392154
View Documents
Bio-Techne
NBP3-09943-100UL
100 μg
459.00€
100µL
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

SF3A2 Polyclonal specifically detects SF3A2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SF3A2
Western Blot
Unconjugated
Rabbit
Human
pre-mRNA splicing factor SF3A, subunit 2, PRP11, PRPF11, SAP 62, SAP62Prp11, SF3a66splicing factor 3a, subunit 2, 66kD, spliceosome associated protein 62, Spliceosome-associated protein 62, splicing factor 3A subunit 2, splicing factor 3a, subunit 2, 66kDa
The immunogen is a synthetic peptide directed towards the N-terminal region of Human SF3A2. Peptide sequence GGKTGSGGVASSSESNRDRRERLRQLALETIDINKDPYFMKNHLGSYECK
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
8175
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.