missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | SFRS4 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18333917
|
Bio-Techne
NBP3-03309-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18392206
|
Bio-Techne
NBP3-03309-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
SFRS4 Polyclonal specifically detects SFRS4 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SFRS4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
6429 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Pre-mRNA-splicing factor SRP75, serine/arginine-rich splicing factor 4, SFRS4, Splicing factor, arginine/serine-rich 4SRP001LB, SRP75SR splicing factor 4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human SFRS4 (NP_005617.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title