Learn More
Tocris Bioscience™ SGC 707
Potent and selective allosteric inhibitor of PRMT3
Brand:  Tocris Bioscience™ 5367/50
543.40 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18759917
  • 613.00€ / 50mg
Estimated Shipment: 09-12-2024
Description
SGC 707 is a potent allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 31 nM). Inhibits PRMT3-mediated methyltransferase activity in vitro. Displays >100-fold selectivity against 31 other methyltransferases and 250 other non-epigenetic targets. Decreases H4R3me2a levels in HEK293 cells in vitro.
Negative control also available.
Specifications
Specifications
>98%
SGC 707
SGC 707
50 mg
298.34