missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3PXD2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18976-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to SH3PXD2A.
Adapter protein involved in invadopodia and podosome formation, extracellular matrix degradation and invasiveness of some cancer cells. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. In association with ADAM12, mediates the neurotoxic effect of amyloid-beta peptide.Specifications
SH3PXD2A | |
Polyclonal | |
Unconjugated | |
SH3PXD2A | |
Adapter protein TKS5, FISH, KIAA0418, SH3 and PX domains 2A, SH3 multiple domains protein 1, SH3MD1, SH3PXD2A, TSK5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14218, 309460, 9644 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O89032, Q5TCZ1 | |
SH3PXD2A | |
Peptide | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |