Learn More
Epredia™ Double Cytoslide™ Microscope Slides
Use Epredia™ Shandon™ Double Cytoslides™ with Epredia™ Shandon™ EZ Double Cytofunnel™ Chambers.
Brand: Epredia™ 5991055
81.53 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Specimen is displayed within a circle marked as 1 or 2
- Number circles correspond with the Cytofunnel™ chamber
Ordering Alert:
- Sold in boxes of 100 slides
Specifications
Glass | |
Double Cytoslide | |
Plain | |
Coated |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
12Hours
4Minutes
26Seconds
27 -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
87Hours
4Minutes
26Seconds
27New Offer Save Up to 85% -
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now