Learn More
Epredia™ Embedding Tissue Cassettes
Hold and identify tissue samples in processing, embedding, and sectioning procedures with Epredia™ Embedding Tissue Cassettes.
76.75€
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
15782482
|
Epredia™
B1003500BL |
Blue | N/A | ||||||
Description
- Designed to accept metal lids
- Designed with round hole pores
- Supplied in boxes of 500 cassettes
Specifications
Round Holes | |
Tissue Cassette |
Product Title
Epredia™ Embedding Tissue Cassettes