Learn More
Sheep anti-Guinea Pig IgA Secondary Antibody, HRP, Invitrogen™
Sheep Polyclonal Secondary Antibody
Brand:  Invitrogen SA510320
Product Code. 17149715
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
This antibody may cross react with IgA from other species
Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Guinea Pig IgA
Polyclonal
HRP
Sheep
Antigen affinity chromatography
RUO
Guinea Pig
Liquid
ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot
1.0 mg/mL
50mM HEPES with proprietary stabilizer and 0.002% thimerosal; pH 7.2
Guinea Pig IgA
0.1 mg
Secondary
4° C
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sheep anti-Guinea Pig IgA Secondary Antibody, HRP, Invitrogen™ > 0.1 mg; HRP