Learn More
Sheep anti-Mouse IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Bio-Techne NB120-6807
Product Code. 18159449
  • 228.00€ / 1mg
Explore more special offers
Description
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Western Blot 1:1000 - 1:5000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:1000 - 1:2000
Mouse IgG whole molecule
1 mg
Adaptive Immunity, Immunology
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Sheep Serum, Mouse IgG and Mouse Serum.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
Sheep
Multi-step
RUO
Secondary
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sheep anti-Mouse IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™ > 1mg; Biotin