Learn More
Sheep anti-Mouse IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Bio-Techne NB120-6807
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
IgG (H+L) | |
Polyclonal | |
Western Blot 1:1000 - 1:5000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:1000 - 1:2000 | |
Mouse IgG whole molecule | |
1 mg | |
Adaptive Immunity, Immunology | |
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Sheep Serum, Mouse IgG and Mouse Serum. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
Sheep | |
Multi-step | |
RUO | |
Secondary | |
Mouse | |
Purified |
Product Content Correction
Product Title
Sheep anti-Mouse IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™ > 1mg; Biotin