missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Rabbit IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9412.1mg
This item is not returnable.
View return policy
Specifications
Sheep anti-Rabbit IgG whole molecule | |
Polyclonal | |
Sheep anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Rabbit IgG whole molecule | |
1 mg | |
No reaction was observed against bovine, chicken, goat, Guinea pig, horse, human, mouse, rat and sheep serum proteins. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Sheep | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Rabbit | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sheep anti-Rabbit IgG whole molecule,Polyclonal Secondary Antibody, Abnova™ > 1mg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction