Learn More
Scotch-Brite™ Sienna Diamond Floor Pads
For cleaning and conditioning worn or dull marble, terrazzo, slate, and polished concrete flooring. When floor appearance reaches a medium shine, the Scotch Brite™ Purple Diamond Floor Pad Plus can be used to further enhance the floor's appearance.
Brand: Scotch-Brite™ 7100200176
This item is not returnable.
View return policy
Description
- Simultaneously cleans and polishes flooring to a brilliant gloss finish without the use of chemicals
- Ideal for worn or dull marble, terrazzo, vinyl, slate, and polished concrete floors
- Easy to use with no expensive or harmful chemicals needed, only water
- Recommended for use with low and standard speed machines
Specifications
Floor Pad | |
505 mm | |
Low and Standard Speed Machines |
Brown | |
Burnishing | |
Round |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Scotch-Brite™ Sienna Diamond Floor Pads > 505 mm