missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-5/CD170 Antibody (07), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06442PE
This item is not returnable.
View return policy
Description
Siglec-5/CD170 Monoclonal antibody specifically detects Siglec-5/CD170 in Human samples. It is validated for Flow CytometrySpecifications
Siglec-5/CD170 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
07 | |
Flow Cytometry | |
CD170, CD170 antigen, CD33 antigen-like 2, CD33L2OB binding protein-2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, SIGLEC-5 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Siglec-5/CD170 (O15389; Met1-Thr 434). | |
0.1 mL | |
Immunology | |
8778 | |
Store at 4°C in the dark. | |
IgG |