Learn More
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography, Fisher Chemical™
Brand:  Fisher Chemical S/0693/60
71.13 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 10403652
  • 123.00€ / 1kg
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Description
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography
Specifications
Specifications
Plastic Powder Jar
1 kg
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
Product Content Correction
Product Title
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography, Fisher Chemical™ > 1kg, Plastic powder jar