Learn More
Silica Gel Grade 923 Davisil™, for Column Chromatography, Fisher Chemical™
Brand: Fisher Chemical S/0697/53
This item is not returnable.
View return policy
Specifications
Plastic Powder Jar | |
5.5 to 8 | |
500 g |
Safety and Handling
Product Identifier
- Silica gel
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%
Product Content Correction
Product Title
Silica Gel Grade 923 Davisil™, for Column Chromatography, Fisher Chemical™ > 500g, Plastic powder jar