Learn More
Borer Chemie™ Silicone-free Grease Tube
Standard quality for desiccators, ground joints, stopcocks and apparatuses. Borer Chemie™ Silicone-Free Grease Tubes are suitable for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metal.
Brand: Borer Chemie™ DGLB0060
This item is not returnable.
View return policy
Description
- Excellent thermal stability
- Available in standard quality (vacuum tight up to 300°C) and high-vacuum quality (up to 400°C)
- Constant lubricating consistency between -40°C and +250°C
- No dripping point
- Chemically stable against most acids, alkalis and gases
- Insoluble in water and low molecular mono- and polyhydric alcohols
Specifications
High-Vacuum | |
60 g |