Learn More
Saint-Gobain Silicone Iso-Versinic™ Stopper
Saint Gobain Performance Plastics™ Silicone Iso-Versinic™ Stoppers have high chemical resistance especially to hydrocarbons, oils, strong acids and aromatic and chlorinated solvents.
Brand:  Saint-Gobain 263008-50
 View more versions of this product
Product Code. 10367934
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Withstands temperatures between -20° and 200°C (possible peaks to 200°C)
  • High chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For collar and tube
4 mm Bottom Diameter
4 mm
Silicone
20 mm
–20 to 200°C
8 mm
ISO-Versinic™ Stoppers