accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Maintenance Lubricants and Penetrating Fluids
VACUUBRAND™ Silicone Oil
10278661
Print
VACUUBRAND™ Silicone Oil
Volume: 2L
Brand:
VACUUBRAND™ 20687501
View more versions of this product
Product Code.
10278661
1460.00€
/ 1 L
Quantity
Estimated Shipment: 02-01-2025
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Product Type
Oil
Quantity
1 Ea.
Volume (Metric)
2 L