accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Maintenance Lubricants and Penetrating Fluids
VACUUBRAND™ Silicone Oil
Print
VACUUBRAND™ Silicone Oil
1460.00€
Specifications
Product Type
Oil
Quantity
1 Ea.
View More Specs
Products
1
Documents
Product Certifications
Products
1
Product Code
Brand
Volume (Metric)
Price
Quantity & Availability
Product Code
Brand
Volume (Metric)
Price
Quantity & Availability
10278661
View Documents
missing translation for 'viewProductCertificates'
VACUUBRAND™
20687501
2 L
1460.00€
1 L
Estimated Shipment: 02-01-2025
Log in
Specifications
Product Type
Oil
Quantity
1 Ea.
Documents
Product Certifications
Back to Top