Learn More
Cole Parmer Silicone Sanitary Gasket, Tri-Clamp
Cole Parmer™ Silicone Sanitary Gasket is designed for Tri-Clamp™ style fittings and comes in a wide variety of materials to match specific process needs.
27.20€
Specifications
Certifications/Compliance | FDA |
---|---|
Material | Silicone |
Quantity | 10/Pk. |
Sterility | Non-sterile |
Type | Gasket |
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | ||||||
11770029
|
Cole Parmer
30548-00 |
3/4 in. |
27.20€
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
Cole Parmer™ Silicone Sanitary Gasket is designed for Tri-Clamp™ style fittings and comes in a wide variety of materials to match specific process needs.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- All gaskets meet USDA, 3-A and CGMP criteria
- FDA Compliant
- All silicone, EPDM, PTFE, and Viton™ gaskets are animal-derived ingredient free and meet USP Class IV.
Specifications
FDA | |
10/Pk. | |
Gasket |
Silicone | |
Non-sterile |
Product Title
Cole Parmer Silicone Sanitary Gasket, Tri-Clamp