Learn More
BRAND™ Silicone Tubing
Translucent tubing
44.85€ - 203.00€
Specifications
Material Silicone Rubber
Overall Length (Metric) 25 m
Color Translucent
View More Specs
Products 5
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
10121681
View Documents
BRAND™
143352
4 mm
44.85€
25m
In Stock
Add to basket
15291389
View Documents
BRAND™
143355
5 mm
52.70€
25m
Estimated Shipment: 03-06-2025
Add to basket
15201399
View Documents
BRAND™
143356
6 mm
61.50€
25m
Estimated Shipment: 06-06-2025
Add to basket
10674162
missing translation for 'viewDocuments'
BRAND™
143359
9 mm
121.00€
25m
In Stock
Add to basket
15251399
missing translation for 'viewDocuments'
BRAND™
143362
12 mm
203.00€
25m
Estimated Shipment: 03-06-2025
Add to basket
Description
Description
  • Autoclavable (121 °C)
  • Note: limited chemical resistance when used with concentrated acids
  • Specifications
    Specifications
    Silicone Rubber
    25 m
    Translucent
    Product Title
    BRAND™ Silicone Tubing