missing translation for 'onlineSavingsMsg'
Learn More

Silver Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical

Product Code. 10181760
Click to view available options
Quantity:
100 mL
Packaging:
HDPE plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7761-88-8
Packaging HDPE plastic bottle
Quantity 100 mL
Color Colorless
Molecular Formula AgNO3
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver solution 10 000 ppm, standard metal solution in approx. 1M nitric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'