Learn More
Bollé Safety SLAM+ Smoke Safety Glasses
Description
Specifications
Specifications
Thickness (Metric) | 2.3 mm |
Lens Coating | Anti-fog, Scratch-resistant |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate, TPR |
UV Protection | Platinum |
Frame Type | Rimless |