Learn More
Kartell Slide Storing System
Slide storing system designed for the classification and storage of microscope slides, made of high impact PMMA. The semi-transparent ice color gives an attractive appearance while the metal structure makes it stable, while ensuring the value of lightness, functionality, strength and durability.
Brand: Kartell 00945L1
Description
- Each single drawer can hold up to 13 supports for slides (code 921) and transparent cover plates (code 920), to be ordered separately
- Each slides support can contain 100 microscope slides, easily extractable
- Each cavity is numbered
- The transparent lids guarantee an easy check of the contents, protecting the slides from dust and other foreign bodies
- The complete Storing System can therefore contain up to 5,200 slide
Specifications
Slide Storage Cabinet | |
5000 | |
Classification and storage of microscope slides | |
4 Swivel Castors |