missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SLU-PP-332

Potent pan-ERR agonist

119.00€ - 387.00€

Specifications

Chemical Name or Material (E)-4-Hydroxy-N'-(naphthalen-2-ylmethylene)benzohydrazide
CAS 303760-60-3
Target Estrogen and Related Receptor Agonists
Molecular Formula C18H14N2O2
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18774384
View Documents
Tocris Bioscience™
8112/50
50 mg
387.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
18724394
View Documents
Tocris Bioscience™
8112/10
10 mg
119.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLU-PP-332 is a potent and selective pan estrogen related receptor (ERR) agonist (EC50 values are 98 nM, 230 nM and 430 nM for ERRα, ERRβ and ERRγ respectively). In a mouse model of metabolic syndrome SLU-PP-332 increases energy expenditure and fatty acid oxidation, decreases fat mass, and improves insulin sensitivity. Also increases mitochondrial respiration in vitro.
Specifications

Specifications

(E)-4-Hydroxy-N'-(naphthalen-2-ylmethylene)benzohydrazide
Estrogen and Related Receptor Agonists
0.98
303760-60-3
C18H14N2O2
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tocris Bioscience™ SLU-PP-332

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.