missing translation for 'onlineSavingsMsg'
Learn More

Smallpox Virus A27L Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA5144503

Product Code. 17950373

  • 355.00€ / 100µL
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Smallpox Virus A27L is a structural protein involved in the envelopment of mature virion (MV) to form the wrapped virion (WV). The wrapping consists of the addition of Golgi membranes to the mature virion. Participates in mature virion (MV) movement within the infected cell. May play an indirect role in MV-cell fusion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Smallpox Virus A27L
Polyclonal
Unconjugated
VARVgp134
Recombinant Vaccinia virus 14 kDa fusion protein (A27L) (1-110AA)
100 μL
Primary
-20°C or -80°C if preferred
IgG
ELISA, Western Blot
1.7 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Rabbit
Protein A/G
RUO
Virus
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Smallpox Virus A27L Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.