Learn More
SNOG, Tocris Bioscience™
Product Code. 18055952
Click to view available options
Quantity:
10 mg
50 mg
NO carrier. Breaks down to release NO
A carrier of NO, relaxing smooth muscle and inhibiting platelet aggregation.
Chemical Identifiers
CAS 57564-91-7
Molecular Formula C10H16N4O7S
Molecular Weight (g/mol) 336.319
InChI Key HYHSBSXUHZOYLX-WDSKDSINSA-N
Synonym s-nitrosoglutathione, gsno, snog, nitrosoglutathione, glutathione thionitrite, n-n-l-gamma-glutamyl-s-nitroso-l-cysteinyl glycine, s-nitroso-l-glutathione, ccris 2095, s-2-amino-5-r-1-carboxymethyl amino-3-nitrosothio-1-oxopropan-2-yl amino-5-oxopentanoic acid, unii-78rri89zto
PubChem CID 104858
ChEBI CHEBI:50091
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-nitrososulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CSN=O)C(=O)NCC(=O)O)C(C(=O)O)N
Specifications
Quantity 50 mg
Formula Weight 336.32
Percent Purity >98%
Chemical Name or Material SNOG