missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPA1 Rabbit anti-Human, Mouse, Rat, Clone: 2D7M1, Novus Biologicals™
Rabbit Monoclonal Antibody
196.00€ - 494.00€
Specifications
Antigen | SNRPA1 |
---|---|
Clone | 2D7M1 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392485
|
Bio-Techne
NBP3-15292-20UL |
20 μg |
196.00€
20µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18348641
|
Bio-Techne
NBP3-15292-100UL |
100 μg |
494.00€
100µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
SNRPA1 Monoclonal antibody specifically detects SNRPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SNRPA1 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Lea1, small nuclear ribonucleoprotein polypeptide A', U2 small nuclear ribonucleoprotein A', U2 small nuclear ribonucleoprotein polypeptide A', U2 snRNP A' | |
A synthetic peptide corresponding to a sequence within amino acids 156-255 of human SNRPA1 (P09661). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
2D7M1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6627 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title