Learn More
Lenz Laborglasinstrumente™ Soda Lime Glass Beads
39.11€ - 52.36€
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
10641672
|
Lenz Laborglasinstrumente™
05124003 |
3mm |
39.11€
1kg |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
10191371
|
Lenz Laborglasinstrumente™
05124002 |
2mm |
39.11€
1kg |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
10101381
|
Lenz Laborglasinstrumente™
05124005 |
5mm |
50.93€
1kg |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
10050191
|
Lenz Laborglasinstrumente™
05124004 |
4mm |
45.53€
1kg |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
10244051
|
Lenz Laborglasinstrumente™
05124006 |
6mm |
52.36€
1kg |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
Specifications
Beads | |
Soda Lime Glass | |
1Pack |