Learn More
BRAND™ Soda Lime Glass Petri Dishes
Perfect for microbial or cell culture applications, these Soda Lime Glass Petri Dishes feature excellent glass quality and workmanship.
Brand: BRAND™ 455701
27.40 EUR valid until 2025-12-31
Use promo code "24345" to get your promotional price.
Description
- Dish and lid are flat inside and outside
- Free from bubbles and streaks
- Cut edges are fire-polished
- Suitable for short-term chemical exposure
Specifications
Soda Lime Glass | |
Petri Dish | |
12 mm |
Round | |
Fire-polished cut edges | |
40 mm |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now