Learn More
BRAND™ Soda Lime Glass Petri Dishes
Perfect for microbial or cell culture applications, these Soda Lime Glass Petri Dishes feature excellent glass quality and workmanship.
Brand: BRAND™ 455717
22.90 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Dish and lid are flat inside and outside
- Free from bubbles and streaks
- Cut edges are fire-polished
- Suitable for short-term chemical exposure
Specifications
Soda Lime Glass | |
Petri Dish | |
15 mm |
Round | |
Fire-polished cut edges | |
60 mm |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
38Hours
3Minutes
27Seconds
15 -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
52Hours
3Minutes
27Seconds
15