missing translation for 'onlineSavingsMsg'
Learn More
Sodium 4-aminosalicylate dihydrate, 98%, Thermo Scientific Chemicals Product Code.: 15496389

Sodium 4-aminosalicylate dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 15496389
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15496389

Brand: Thermo Scientific Alfa Aesar J64449.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-19-5
Molecular Formula C7H10NNaO5
Molecular Weight (g/mol) 211.149
MDL Number MFCD00151044
InChI Key GMUQJDAYXZXBOT-UHFFFAOYSA-M
Synonym sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium uspShow MoreShow Less
PubChem CID 16211148
IUPAC Name sodium;4-amino-2-hydroxybenzoate;dihydrate
SMILES C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+]

Specifications

Melting Point 250°C
Color White
Quantity 100 g
Sensitivity Air and light sensitive
Formula Weight 211.14 (175.12ahy)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sodium 4-aminosalicylate dihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM03L030 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberJ64449.22,J64449.14,J64449.36
Lot NumberM13G009 Certificate TypeCertificate of Analysis Date03/06/2022 Catalog NumberJ64449.22,J64449.14,J64449.36
Lot NumberY08E049 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64449.22,J64449.14,J64449.36
Lot NumberY05E060 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64449.22,J64449.14,J64449.36
Lot NumberI28Y030 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ64449.22,J64449.14,J64449.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium 4-aminosalicylate dihydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.