Learn More
Sodium AAS Standard, 1000 mg/L Na in water, Honeywell Fluka™
Brand:  Honeywell Fluka E4050-100ML
 View more versions of this product
Product Code. 16624354
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sodium AAS Standard
Standards and Modifiers for AAS
0.999 g/cm3
100 mL