Learn More
Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.0 Product Code.: 15442909
Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.0
Product Code. 15442909
250 mL
Click to view available options
Quantity:
250 mL
500 mL
Product Code. 15442909
Brand: Thermo Scientific Chemicals J60066.AK
Request Bulk or Custom Format
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O
Specifications
Concentration 1 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Sodium bicarbonate buffer soln.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR01L563 Certificate TypeCertificate of Analysis Date11/05/2025 Catalog NumberJ60066.AK
Lot NumberR01L564 Certificate TypeCertificate of Analysis Date11/05/2025 Catalog NumberJ60066.AK
Lot NumberZ14K549 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberJ60066.AK,J60066.AP
Lot NumberM11L573 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog NumberJ60066.AK,J60066.AP
Lot NumberX23K538 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberJ60066.AK,J60066.AP
Product Title
Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.0 >