missing translation for 'onlineSavingsMsg'
Learn More
Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate, Fisher Chemical™ Product Code.: 10457840

Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate, Fisher Chemical™

Product Code. 10457840 Shop All Fisher Chemical Products
500 g, 500g
Click to view available options
Quantity:
500 g
3 kg
Unit Size:
3kg
500g
This item is not returnable. View return policy

Product Code. 10457840

Brand: Fisher Chemical S/P520/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.006
MDL Number 149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]

Specifications

Melting Point 60°C
Color White
pH 4.5
Physical Form Solid
Quantity 500 g
Formula Weight 156.01g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.