Learn More
Sodium diphenylamine-4-sulfonate, ACS, Thermo Scientific Chemicals
Redox indicator
46.63€ - 337.68€
Chemical Identifiers
CAS | 6152-67-6 |
---|---|
Molecular Formula | C12H11NNaO3S |
Molecular Weight (g/mol) | 272.27 |
MDL Number | MFCD00036468 |
InChI Key | VMVSDVIYIMNZBS-UHFFFAOYSA-N |
Synonym | sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium saltShow More |
PubChem CID | 517002 |
IUPAC Name | sodium;4-anilinobenzenesulfonate |
SMILES | [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348858
|
Thermo Scientific Alfa Aesar
041714.04 |
2 g |
46.63€
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338858
|
Thermo Scientific Alfa Aesar
041714.09 |
10 g |
141.71€
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358858
|
Thermo Scientific Alfa Aesar
041714.18 |
50 g |
337.68€
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Redox indicatorSodium diphenylamine-4-sulfonate is used as a pH independent redox indicator used in the determination of redox potential. It is used in the preparation of poly-(aninile-co-N-(4-sulfophenyl)aniline) by polymerization with aniline. It acts as a ligand and form complexes with metal halides like lanthanum(III) chloride. It is also involved in the synthesis of gold nano crystals by reaction with chloroauric acid in the presence of polyethylene glycol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6152-67-6 | |
272.27 | |
VMVSDVIYIMNZBS-UHFFFAOYSA-N | |
517002 | |
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 |
C12H11NNaO3S | |
MFCD00036468 | |
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium saltShow More | |
sodium;4-anilinobenzenesulfonate |
Specifications
6152-67-6 | |
MFCD00036468 | |
Soluble in water. | |
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 | |
272.27 | |
271.27 | |
Powder |
C12H11NNaO3S | |
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt | |
VMVSDVIYIMNZBS-UHFFFAOYSA-N | |
sodium;4-anilinobenzenesulfonate | |
517002 | |
ACS | |
Sodium diphenylamine-4-sulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 228-165-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.