Learn More
Sodium Dodecyl Sulfate (SDS), ≥95%, Thermo Scientific Chemicals Product Code.: 15875308
Sodium Dodecyl Sulfate (SDS), ≥95%, Thermo Scientific Chemicals
Product Code. 15875308 Shop All Thermo Scientific Products
5 kg, Cardboard Carton
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Cardboard Carton
Plastic bottle
Product Code. 15875308
Brand: Thermo Scientific J18220.A7
Request Bulk or Custom Format
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Chemical Identifiers
CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate,n to Dodecyl sulfate sodium salt
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Specifications
Physical Form Crystalline Powder
Melting Point ∼206°C
Quantity 5 kg
Packaging Cardboard Carton
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
IUPAC Name sodium;dodecyl sulfate
Formula Weight 288.38
Percent Purity ≥95%
Chemical Name or Material Sodium Dodecyl Sulfate (SDS)
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number232215 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number242192 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number200082 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number231190 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number232201 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberJ18220.A1,J18220.36,J18220.A7
compliance-icons compliance-icons
Product Identifier
  • Sodium dodecyl sulfate, Thermo Scientific
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulfate
Product Title
Sodium Dodecyl Sulfate (SDS), ≥95%, Thermo Scientific Chemicals >