Learn More
Sodium Hydroxide, 0.1M, Chem Lab™
Click to view available options
Quantity:
0.5 L
10 L
Specifications
Specifications
pH | >11 |
Color | Colorless |
Concentration | 4g NaOH/l H2O = 0.1 N (±0.0002/20°C) |
Density | 1.01g/mL |
Solubility Information | Soluble in Water |
Quantity | 0.5 L |
Odor | Odorless |
Physical Form | Liquid |