Learn More
Tocris Bioscience™ Sodium orthovanadate
Protein tyrosine phosphatase inhibitor
Brand:  Tocris Bioscience™ 2821/100
Product Code. 18742774
  • 129.00€ / 100mg
Estimated Shipment: 30-01-2025
Description
Sodium orthovanadate is an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and ATPases. Suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.
Specifications
Specifications
100 mg
Sodium orthovanadate
183.91
Sodium orthovanadate