Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.6
83.52€ - 132.66€
Specifications
Chemical Name or Material | Sodium phosphate buffer soln. |
---|---|
Concentration | 0.5 M |
pH | 7.6 |
Physical Form | Liquid |
MDL Number | MFCD00146206 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15432979
|
Thermo Scientific Chemicals
J60158.AK |
250 mL |
83.52€
250mL |
In Stock
Log in to see stock. |
|||||
15442979
|
Thermo Scientific Chemicals
J60158.AP |
500 mL |
132.66€
500mL |
In Stock
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium phosphate buffer soln. | |
7.6 | |
MFCD00146206 |
0.5 M | |
Liquid |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures