missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The thiosulfate anion reacts stoichiometrically with iodine in aqueous solution. Due to the quantitative nature of this reaction, sodium thiosulfate is used as a redox titrant in volumetric analysis and is a component of iodine clock experiments. It can be used to measure the oxygen content of water and to estimate the concentration of certain compounds in solution (hydrogen peroxide, chlorine). It also has many medicinal uses.
Specifications
Specifications
Boiling Point | 100°C |
Melting Point | 48.3°C |
Physical Form | Liquid |
Quantity | 10 L |
Solubility Information | Soluble in water |
Formula Weight | 248.18g/mol |
Concentration or Composition (by Analyte or Components) | 0.1mol/L |
Density | 1.00g/mL |
Chemical Name or Material | Sodium Thiosulfate Pentahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Thiosulfate, 0.1M, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction