Learn More
Thermo Scientific™ SoftFit-L™ Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
Brand: Thermo Scientific™ 3704-ER
27.36 EUR valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
Specifications
Not Autoclavable | |
Empty Rack for 20 ÎĽL, 200 ÎĽL and 300 ÎĽL tip inserts | |
Pipet Tip Rack | |
ART SoftFit |
SoftFit L Rack | |
10 Racks/Pk., 5 Pks./Cs. | |
20 ÎĽL Pipet Tip, 200 ÎĽL Pipet Tip, 300 ÎĽL Pipet Tip |
Promotions
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30