Learn More
Memmert™ Software AtmoCONTROL for UF55plus
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand:  Memmert™ UF55plusFDAQ1
Product Code. 17569505
  • 2625.00€ / Each
Estimated Shipment: 02-07-2025
Add to basket
Description
Description
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
UF55plus Oven
Software
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Software AtmoCONTROL for UF55plus > For Use With: UF55plus