Learn More
Zeiss™ Software - ZEN 3.3 Lite
ZEN lite has reduced basic functionality and user interface of ZEN pro, desk or system. Drivers for Axiocam cameras, image acquisition (SNAPs) and viewer, no microscope control.
Brand: Zeiss™ 410135-1001-330
This item is not returnable.
View return policy
Description
- ZEN lite is based on the powerful ZEN microscopy software in both looks and functions
- However, unlike the full versions – ZEN desk, ZEN system and ZEN pro – its functionality is only basic
- ZEN lite does not need a dongle or a license.
- A number of selected, paid-for modules are available to expand it.
Specifications
Modules |