Learn More
Brannan™ Soil Analysis pH Meter
Instantly tells you how acid or alkaline yo soil is and checks which plants suit.
Brand:  Brannan™ 13/716/0
Product Code. 13100443
  • 38.60€ / Each
Estimated Shipment: 19-03-2025
Description
  • Includes a plant care booklet listing over 400 pH levels
Specifications
Specifications
Cable Probe
3.5 to 9
Removable
6.8 cm
Soil Analysis
0.085 kg
pH Meter
10 cm
Product Title
Brannan™ Soil Analysis pH Meter > Probe Type: Removable