Learn More
Cole-Parmer™ Soil Sampling Handle
Brand: Cole-Parmer™ 99025-02
This item is not returnable.
View return policy
Specifications
Soil Sampling Handle | |
Nickel-plated Carbon Steel | |
Five Different Sampling Handles | |
304 to 457.2 mm |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Cole-Parmer™ Soil Sampling Handle > Material: Nickel-plated carbon steel; Sampling Depth: 30 to 46cm; Includes: Five Different Sampling Handles