Learn More
Gilac™ Solid Container with Attached Lid
Solid Container with Attached Lid has a hinge-attached lid to better protect contents with no risk of losing the lid. An ideal container for transporting tableware.
Brand: Gilac™ G121003
Additional Details : Weight : 3.03000kg
Description
- Solid container with hinge-attached lid to better protect contents.
- No risk of losing the lid.
- Ideal container for transporting tableware.
- Designed with smooth sides for easy cleaning.
- Ergonomic integrated handles on all four sides.
- Stackable container with vertical reinforcement for greater durability.
- Compatible with 600 x 400 dollies for easy transport.
- Easy-to-seal lids with cord to prevent theft.
Specifications
Storage Tote | |
Gray | |
11.9 gal. | |
Hinge-attached Lid |
Polypropylene | |
Rectangular | |
45 L | |
600 x 400 x 246 mm |