Learn More
Fisherbrand™ Solid Neoprene Rubber Bungs
Used for laboratory purposes.
Brand:  Fisherbrand™ NB006/S
 View more versions of this product
Product Code. 11507093
  • 5.00€ / Pack of 20
Only null left
Estimated Shipment: 04-07-2025
Add to basket
Description
Description
  • Made up of neoprene rubber
  • Hardness is 46/55 IRHD
Specifications
Specifications
For laboratory use
16 mm
Gray
8 mm
Bung
46/55 IRHD
20/Pk.
6 mm
Neoprene
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Solid Neoprene Rubber Bungs > Bottom Diameter: 6mm