Learn More
Fisherbrand™ Sound Protection Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Reduces the noise generated by ultrasonic cleaning units while providing a secure closure. For use with Ultrasonic Water Baths.
380.00€ - 549.00€
Specifications
Type | Sound Protection Cover |
---|---|
Style | Hinged Lid |
For Use With (Application) | Noise reduction for ultrasonic cleaning units |
Material | Stainless Steel |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
15626074
|
Fisherbrand™
1076202 |
Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H |
549.00€
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
15636074
|
Fisherbrand™
1076206 |
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H |
499.00€
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
15646074
|
Fisherbrand™
1076211 |
Ultrasonic Cleaner, Elmasonic Xtra ST 600 H/800 H |
394.00€
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||||
15656074
|
Fisherbrand™
1076213 |
Ultrasonic Cleaner, Elmastonic Xtra ST 300 H/500 H |
380.00€
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
Description
- Hinged design
- Available in a range of sizes for all X-tra ST models
- Manufactured from stainless steel
- Accessory item
Specifications
Sound Protection Cover | |
Hinged Lid | |
Noise reduction for ultrasonic cleaning units | |
Stainless Steel |
Product Title
Fisherbrand™ Sound Protection Cover for Elmasonic X-tra ST Ultrasonic Cleaners