missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blue Eagle Spare visors for K-Series Safety Face Shields
Click to view available options
Product Type: Visor
Description
- Visor made of 0.8 mm thick polycarbonate plastic
- Tested and approved for broader temperature working environment
- Spare for use with Blue Eagle K-Series Safety Face Shields with Browguard and Visor (Catalog # 16609893, 16622113, 16689883, and 16699883
- Approved for use in a wider temperature range and are available in different colors for convenience.
Compliance
This product complies with the essential requirements for health and safety as they are provided by the European Regulation (EU) 2016/425 for Personal Protective Equipment.
Specifications
Specifications
Thickness (Metric) | 0.8 mm |
Color | Clear |
Product Type | Visor |
For Use With (Equipment) | K-Series Safety Face Shields |
Material | Polycarbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'